Would you like to log in again?
Steraffirm™ VH₂O₂ Process Indicator, STERIS®
Supplier: Steris
Steraffirm™
PCC049
MSPP-PCC049EA 93.85 USD InStock
MSPP-PCC049
Steraffirm™ VH₂O₂ Process Indicator, STERIS®
Sterilization Indicators
Steraffirm VH₂O₂ process indicator is a type 1 hydrogen peroxide vapor chemical indicator.
Steraffirm VH₂O₂ process indicator consists of a Tyvek®1 strip. The process indicator has a magenta indicator ink circle and a yellow endpoint color standard circle. The magenta indicator ink circle is specially formulated to be responsive to hydrogen peroxide vapor during the biodecontamination cycle and exhibits a gradient color change from magenta to orange to yellow. The yellow endpoint color standard circle is used as a visual comparison to the indicator ink.
Certifications: Meets ISO 11140-1:2014.
Delivery information: Steraffirm process indicator includes instructions for use (IFU). Package label, indicator strip and adhesive label include lot number, expiration date and reorder number.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty