Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fetal Bovine Serum, Charcoal-Dextran Treated, Omega Scientific

Supplier: Omega Scientific
FB-04
MSPP-FB04EA 1069.23 USD InStock
MSPP-FB04
Fetal Bovine Serum, Charcoal-Dextran Treated, Omega Scientific
Sera (Serum)
Charcoal dextran-treated fetal bovine serum has lower levels of hormones and steroids.

  • Virus screened
  • Mycoplasma tested
  • Store at –20 °C
  • Triple 0.1 µm filtered
  • Weight: 1.5 lbs.
  • Dimensions: 2.91339×2.91339×6.9685"

The concentration of these are reduced using charcoal, dextran and NaCl until progesterone concentration is less than 0.2 ng/ml.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty