Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dodecyl sulfate (SDS) ≥99%, Molecular Biology Grade, Calbiochem®, Millipore®

Supplier: MilliporeSigma
Danger
Synonyms: Sodium dodecyl sulfateLauryl sulfate sodium saltSodium lauryl sulfateDodecyl sulfate sodium saltDodecyl sulphuric acid sodium saltLauryl sulphate sodium saltSodium lauryl sulphateDodecyl sulphate sodium saltDodecyl sulfuric acid sodium salt

428023-500GM
80503-356EA 497.02 USD InStock
80503-356
Sodium dodecyl sulfate (SDS) ≥99%, Molecular Biology Grade, Calbiochem®, Millipore®
Sodium dodecyl sulphate (SDS)


Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/54/91/735491.jpg
Order Now
Specification Test Results
Purity ≥99%
DNases, proteases, RNases none detected
Heavy metals ≤0.0005%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty