Would you like to log in again?
Tronex® 9117 Series NEW AGE® Chemo-Rated Nitrile Examination Gloves, Tronex International Inc.
Supplier: Tronex
Tronex®
9117-35 9117-10 9117-20
76324-964BX 71.73 USD InStock
76324-964 76324-978 76324-974
Tronex® 9117 Series NEW AGE® Chemo-Rated Nitrile Examination Gloves, Tronex International Inc.
Gloves
Tronex® nitrile exam-grade gloves feature an ambidextrous fit with beaded cuffs. They are designed to meet or exceed all industry standards for safety and quality.
  • Excellent barrier protection, comfort, and chemical resistance
  • Fingertip textured for improved grip
  • Not made with natural rubber latex, powder-free, non sterile
Tronex® 9117 series examination gloves feature an innovative nitrile material formulation for exceptional fit and feel properties for applications with shorter durations. Gloves offer excellent barrier protection for bloodborne pathogens, as well as resistance to chemotherapy drugs and other chemicals.
Certifications: Chemo-rated per ASTM D6978.
Packaging: Gloves are packaged 250 per box, 10 boxes per case.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty