Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PhysiciansCare® by First Aid Only® First Aid Kit Rubbing Alcohol, Isopropyl Alcohol, Acme United

Supplier: Acme United
PhysiciansCare®
FAOM313
76325-904EA 14.11 USD InStock
76325-904
PhysiciansCare® by First Aid Only® First Aid Kit Rubbing Alcohol, Isopropyl Alcohol, Acme United
First Aid Supplies First Aid Kit Components
First aid antiseptic works to decrease germs in minor cuts and scrapes.

  • For use with all first aid kits
  • Contains 70% isopropyl alcohol to ensure effectiveness
  • Use as stand-alone first aid

Packaging: Packaged in a convenient 16 oz. bottle.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty