Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

KHS-101 ≥98% (by HPLC)

Supplier: BioVision
Synonyms: N4-isobutyl-N2-((2-phenylthiazol-4-yl)methyl)pyrimidine-2,4-diamine

AB287041-25MG AB287041-5MG
76325-700EA 1057.19 USD InStock
76325-700 76325-698
KHS-101 ≥98% (by HPLC)
KHS-101
KHS-101 is a selective inducer of neuronal differentiation. It induces neuronal differentiation in cultured hippocampal neural progenitor cells (NPCs) by interacting with TACC3 (EC50 ~1 μM). Also induces acceleration of neuronal differentiation in the hippocampal dentate gyrus in vivo. Additionally, it disrupts energy metabolism in human glioblastoma cells and reduces tumor growth in mice.

  • Neuronal differentiation inducer


Formula: C₁₈H₂₁N₅S
MW: 339.46 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18206916
CAS Number: 1262770-73-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/87/73/27368773.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty