Would you like to log in again?
1 L Process Filter Reactors, Cylindrical, Unjacketed, Electric, Chemglass
Supplier: Chemglass
CG-1959-U1000
10170-266EA 21101.07 USD InStock
10170-266
1 L Process Filter Reactors, Cylindrical, Unjacketed, Electric, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Unjacketed filter reactors can be used for single or multi-step reactions and filtrations in the same vessel, and large scale peptide synthesis.
Filter base eliminates material transfers and significantly reducing losses. Complete reactor system on our ChemRxnHub stand. Supplied with the components listed in the link below. 150°C max. temp. limit for filter base.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty