Would you like to log in again?
PicoProbe™ Glutamate Carboxypeptidase II Activity Assay Kit, BioVision
Supplier: BioVision
AB284558-100TEST
76326-382EA 993.29 USD InStock
76326-382
PicoProbe™ Glutamate Carboxypeptidase II Activity Assay Kit, BioVision
Assays Enzyme Assays
Sensitive assay for the detection and quantification of active glutamate carboxypeptidase.
  • Sensitive assay for measuring GCPII activity in various biological samples
  • HTS adaptable
  • Simple procedure
  • Fast and convenient
Glutamate carboxypeptidase II [GCPII, EC 3.4.17.21; N-acetylated-alpha-linked acidic dipeptidase (NAALADase), prostate-specific membrane antigen (PSMA) or folate hydrolase (FOLH1)] is a zinc-metallopeptidase, multifunctional protein. In humans, it is mainly expressed in the nervous system, prostate, small intestine and kidney. In males, PSMA is overexpressed in patients suffering prostate cancer and thus, it is used as a diagnostic marker for this disease. NAALADase, the brain isozyme, plays an important role in glutamate biosynthesis and could be used for treating neuronal damage caused by excess glutamate in brain. BioVision’s PicoProbeTM glutamate carboxypeptidase II activity assay kit is a simple plate-based fluorometric assay for the measurement of GCPII activity in biological Samples. The provided substrate is transaminated in the presence of GCPII producing glutamate. The detection system is based on an enzymatic reaction in which a fluorogenic probe is reduced generating a stable signal. The reduced fluorophore produces a strong signal which is directly proportional to the amount of active GCPII in samples. The assay can detect as low as 0.5 µU of glutamate carboxypeptidase II.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty