Would you like to log in again?
Phosphatidylglycerol Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB273288-100TEST
76326-388EA 975.84 USD InStock
76326-388
Phosphatidylglycerol Assay Kit (Fluorometric), BioVision
Assays Cellular Assays
The first kit in the market that detects phosphatidylglycerol levels in biological samples.
  • Can be completed in less than one hour
  • Specific
  • Validated with biological samples
  • Simple and easy protocols
Phospholipids are physiologically critical amphiphilic molecules that are involved in regulation of numerous cellular processes. Phosphatidylglycerol (PG) is a glycerophospholipid consisting of a glycerol backbone with two acyl chains and a phosphatidyl group that carries another glycerol moiety. Lung surfactant is an important lipid-protein mixture that aids in expansion and prevents total collapse of the lung during respiration. PG is a prominent phospholipid in lung surfactant, and its presence can be monitored as a measure of lung development in the maturing fetus. In addition, most bacteria utilize PG in their membrane, and plants utilize this component as the major phospholipid in the photosynthetic membranes of leaf tissue. In HPLC analysis of phospholipids, phosphatidic acid can interfere with and complicate determination of PG content. BioVision’s phosphatidylglycerol Assay Kit utilizes specific enzymes to generate an intermediate that reacts with a probe, yielding a signal that can be quantified fluorometrically, and is proportional to the amount of PG present in the sample, unaffected by the presence of phosphatidic acid, cardiolipin, or other lipids. When used as described, the assay is highly selective against other phospholipids (see figure below) and capable of detecting as little as 20 pmoles of phosphatidylglycerol.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty