Would you like to log in again?
Glucose Isomerase Activity Assay Kit (Colorimetric), BioVision
Supplier: BioVision
AB273289-100TEST
76326-340EA 826.46 USD InStock
76326-340
Glucose Isomerase Activity Assay Kit (Colorimetric), BioVision
Assays Enzyme Assays
Easy kit for detecting and characterizing glucose Isomerase preparations.
  • Simple, rapid and convenient
  • Compatible with bacterial lysates and purified protein/preparations
Glucose (Xylose) isomerase (GI) (EC 5.3.1.5) can catalyze the reversible conversion of glucose to fructose, and xylose to xylulose. GI has important industrial applications in the production of high-fructose corn syrup (HFCS). It is known HFCS is sweeter than glucose; thus it is widely used in foodstuff and pharmaceutical products. Additionally, additional research on using glucose isomerase in the production of biofuels has increased. BioVision’s glucose isomerase activity assay kit provides a quick and easy method for monitoring GI activity in various samples. In this assay, glucose isomerase converts fructose into glucose. This isomerization is detected through a series of enzymatic reactions generating an intermediate that reacts with the probe producing a stable chromophore with strong absorbance at OD = 570 nm. The assay is simple, sensitive and can detect glucose isomerase activity as little as 50 µU in a variety of samples.
Delivery information: Kit includes all the reagents required to measure GI activity
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty