Would you like to log in again?
Ethanolamine Kinase (ETNK) Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB273322-100TEST
76326-342EA 917.44 USD InStock
76326-342
Ethanolamine Kinase (ETNK) Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
The first kit in the market that measures ENTK activity in biological samples.
  • Non-radiometric fluorescent detection
  • HTP compatible
  • Takes 1 to 2 hours
  • Simple protocols/ sample preparations
Ethanolamine kinase (ETNK; E.C. 2.7.1.82), catalyzes the ATP-dependent phosphorylation of free intracellular ethanolamine and is the first committed step of the de novo CDP-ethanolamine pathway for phosphatidylethanolamine (PE) biosynthesis in mammalian tissues. PE is a major phospholipid component of cell membranes and it is involved in diacylglycerol and G protein-dependent signal transduction. ETNK plays a critical role in cell growth and it may be associated with abnormal choline and ethanolamine phospholipid metabolism in breast, prostate, ovarian and cervical cancers. Two isoforms of ethanolamine kinase, ETNK1 and ETNK2, have been identified in mammalian cells and sharing >50% homology. In BioVision’s Ethanolamine Kinase activity assay kit, ENTK phosphorylates ethanolamine producing phosphoethanolamine and ADP. The produced ADP is then detected with a set of enzymatic reactions that generate fluorescencent product (Ex/Em: 535/587 nm). The fluorescence signal is directly proportional to the generated ADP. BioVision’s ETNK assay is suitable for detecting ETNK enzyme activity in different tissues and it can detect as low as 0.1 mU of ETNK in biological samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty