Would you like to log in again?
Viral DNA Extraction and Purification Kit, BioVision
Supplier: BioVision
AB288101-100TEST
76326-354EA 432.2 USD InStock
76326-354
Viral DNA Extraction and Purification Kit, BioVision
Assays Nucleic Acid Assays
Viral DNA extraction and purification.
  • Yields high pure viral DNA
  • Kit can also be used with a variety of automatic nucleic acid extraction instruments or workstations
This kit allows for extraction and purification of viral DNA from serum, plasma, urine, lymph, cell culture supernatants or from a variety of viral-containing fluids. Using proprietary viral lysis solution, viral DNA can be efficiently extracted and then purified using magnetic beads, yielding high pure viral DNA with a ratio of OD260/280 between 1.75 and 1.85. The recovered DNA size can be up to 60kb. The kit will work with a 48 well round bottom plates if a special magnetic frame is used. The kit can also be used with a variety of automatic nucleic acid extraction instruments or workstations.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty