Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PCR-Listeria monocytogenes Detection Kit, BioVision

Supplier: BioVision
AB287886-96TEST
76326-182EA 962.92 USD InStock
76326-182
PCR-Listeria monocytogenes Detection Kit, BioVision
Assays Nucleic Acid Assays
PCR based Listeria monocytogenes detection kit.

  • Inclusivity: Positive in 48 reference strains of L. monocytogenes (serovars 1 to 7) from different collections
  • Reliable and highly specific results
  • Detection: probe labelled with fluorescent dyes Listeria monocytogenes: FAM-BHQ1a; IAC: HEXBHQ1
  • Fast: results in less than 24 hours with minimal handling
  • Quantification limit: 20 gµ per reaction (100%)
  • Quantification dynamic range: 6 logs
  • Exclusivity: 100% tested with 94 nontarget strains composed of 51 strains of Listeria nomonocytogenes and 43 strains of non-listeria
  • Easy and ready-to-use kit
  • Shipping temperature: Dry ice

This PCR-Listeria monocytogenes detection kit is an ideal tool for simple, fast and reliable amplification and detection of specific DNA fragments from Listeria monocytogenes by the real-time PCR method. The Kit includes all reagents required in a comfortable ready-to-use Multiplex PCR MasterMix. The optimized MasterMix contains a buffer, dNTPs, Hot-start DNA polymerase, DNA-free water, MgCl2 and an Internal Amplification Control (IAC) whose detection indicates the absence of PCR inhibitors. Primers and Probes for the amplification of IAC as well as for the amplification of the target gene are included in the MasterMix. The Probe for the detection of target gene is labelled with the FAM, whereas the probe for the detection of IAC is labelled with the HEX fluorochrome. In addition, the kit includes both positive control and negative control. The positive control is supplied to demonstrate that the PCR amplification is working efficiently with the supplied components. To confirm absence of contamination, a negative control reaction should be included every time the kit is used. Includes DNAready Lysis Buffer to extract the DNA from the sample prior to PCR detection.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty