Would you like to log in again?
PicoProbe™ Glutaminase (GLS) Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB284547-100TEST
76326-196EA 978.11 USD InStock
76326-196
PicoProbe™ Glutaminase (GLS) Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
The most sensitive, specific assay for the measurement of GLS activity in biological samples.
  • Validated with relevant biological samples
  • Includes human GLS as positive control
  • Sensitive, specific assay for detection of glutaminase activity in biological samples
Glutaminase (EC 3.5.1.2) is a mitochondrial enzyme that hydrolyzes glutamine producing glutamate and ammonia. GLS is one of the two key enzymes that are responsible for glutamine homeostasis, the other being glutamine synthetase. Since glutamine is a major metabolic substrate involved in gluconeogenesis, its homeostasis is tightly controlled. Glutaminase has tissue-specific roles in multiple organs, with the isoform GLS1 being expressed in kidneys and brain, and GLS2 being expressed in the liver. It is involved in maintenance of acid-base homeostasis by producing ammonia during renal acidosis in kidneys; and regulation of glutamate –which acts as a neurotransmitter- in the brain. On the other hand, GLS 2 regulates the generation of ammonia in liver by producing urea. Research shows GLS increased activity in individuals facing starvation, diabetes and high protein diets. BioVision’s PicoProbeTM glutaminase activity assay kit is a simple plate-based fluorometric assay for measuring glutaminase activity in biological samples. GLS hydrolyzes glutamine forming glutamate and ammonia. Glutamate, in the presence of a developer and enzyme mix converts a non-fluorescent probe to a fluorescent product via an enzymatic reaction. The assay can detect as low as 2 µU of glutaminase in biological samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty