Would you like to log in again?
Glutaminase (GLS1) Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB283389-100TEST
76326-198EA 1205.21 USD InStock
76326-198
Glutaminase (GLS1) Inhibitor Screening Kit (Fluorometric), BioVision
Assays Enzyme Assays
Sensitive, reproducible assay for initial screening of potential glutaminase Inhibitors.
  • Sensitive, specific assay for screening GLS1 inhibitors
  • Includes hrGLS1 and CB-839 (inhibitor control)
Glutaminase (EC 3.5.1.2, GLS) is an enzyme that hydrolyzes glutamine producing glutamate and ammonia. It has tissue-specific roles in multiple organs, some of which include ammonia generation for urea synthesis in the liver; maintenance of acid-base homeostasis by ammonia production during renal acidosis in the kidney; and regulation of neurotransmitter glutamate in the brain. The isoform glutaminase 1 (GLS1) or phosphate-activated mitochondrial glutaminase is critical for glutamine utilization by cancer cells and the rate of glutaminolysis is known to increase in tumors, making it extremely important for tumor cell metabolism. GLS1 inhibitors have been considered as potential candidate drugs for cancer therapy. This glutaminase (GLS1) inhibitor screening kit is a plate-based fluorometric kit for screening human GLS1 inhibitors. The hydrolysis of glutamine by glutaminase forms glutamate and ammonia. Glutamate, in the presence of a developer and enzyme mix, converts a non-fluorescent probe to a fluorescent product via an enzymatic reaction. A well-known glutaminase inhibitor - CB-839 is provided as an inhibitor control.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty