Would you like to log in again?
Aldose Reductase Inhibitor Screening Kit (Colorimetric), BioVision
Supplier: BioVision
AB283360-100TEST
76326-206EA 902.17 USD InStock
76326-206
Aldose Reductase Inhibitor Screening Kit (Colorimetric), BioVision
Assays Enzyme Assays
To screen for potential AR inhibitors.
  • Rapid and simple
  • Sensitive and reliable
Aldose reductase (aldehyde reductase; AR; ALR2; EC 1.1.1.21), a member of the aldo-keto reductase superfamily, catalyzes the NADPH-dependent reduction of a wide variety of aldoses (molecules containing carbonyl groups) to their corresponding alcohols. It is a key enzyme in the polyol pathway and the key factor in the reduction of glucose to sorbitol. Synthesis and accumulation of sorbitol due to AR activity is the main cause of diabetic complications, such as diabetic cataract, retinopathy, neuropathy and nephropathy. Inhibition of AR has been successfully used in clinical settings. These approved drugs prevent the accumulation of sorbitol. Thus, preventing eye and nerve damage in patients with diabetes. In addition, recent studies have shown that AR inhibitors may be able to prevent or delay the onset of cardiovascular complications such as ischemia/reperfusion injury, atherosclerosis and atherothrombosis. BioVision’s aldose reductase inhibitor screening kit can be used to screen potential inhibitors of this molecular target. Epalrestat, a noncompetitive AR inhibitor, is used as a positive control. In this assay, AR activity is monitored by the reduction in absorbance reading at OD 340 nm, while potential inhibitors arrest this decrease. The kit is adapted to a 96-well format and provides a rapid, simple, sensitive and reliable test for screening of AR inhibitors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty