Would you like to log in again?
EZClick™ Stearoylated Protein Assay Kit (FACS/Microscopy), Red Fluorescence, BioVision
Supplier: BioVision
AB273280-100TEST
76326-212EA 826.45 USD InStock
76326-212
EZClick™ Stearoylated Protein Assay Kit (FACS/Microscopy), Red Fluorescence, BioVision
Assays Protein Assays
For labeling and detection of stearoylated proteins.
  • Simple procedure
  • Highly specific
  • Robust method for labeling and detection of stearoylated proteins
Stearoylation is one of the numerous types of post-translational modification in proteins. Stearoylated proteins are involved in the regulation of cellular processes such as apoptosis, growth and metabolism. In this modification Stearic acid, a saturated fatty acid is covalently bound to a protein through the action of long-chain-fatty-acid-[acyl-carrier-protein] ligase. In the presence of ATP, ligase then transfers a stearate moiety to the target protein forming a carbon-sulfur bond (C=S). Recent studies showed a possible correlation between mitochondrial dysfunction and some diseases including neurodegenerative disease, cancer and aging. Furthermore, it is known that stearoylated Transferrin Receptor (TfR1) inhibits JNK activation thereby reducing mitofusin ubiquitination and restoring mitochondrial function. Studying stearoylation of proteins can shed a light on disease etiology and prevention. This EZClick™ stearoylated protein assay kit is a highly specific, simple and robust method for labeling and detection of stearoylated proteins. The kit uses alkynyl stearic acid that is fed directly into the cells and gets incorporated into proteins during or post translation. This modification can be followed by click reaction with an azide-containing dye. The assay kit offers a powerful method for imaging localization, trafficking, and dynamics of stearoylated proteins or detection by FACS for quantitative studies. Labeled stearoylated proteins can be directly detected in 1D or 2D gels using the appropriate excitation sources or enriched by immunoprecipitation with biotin-azide or antibodies prior to proteomic analysis. We provide sufficient materials for 100 assays in a 96-well plate format.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty