Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid 95.0-97.0%, ElectroPUR® SEMI

Supplier: Puritan Products, Inc.
Danger

287785 287775SC 287715 287775 287734
76266-636EA 848.55 USD InStock
76266-636 76266-634 76266-640 76267-186 76266-638
Sulfuric acid 95.0-97.0%, ElectroPUR® SEMI
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Color Max. 10 APHA
Assay (w/w) 95.0 - 97.0%
Chloride Max. 0.1 ppm
Nitrate Max. 0.2 ppm
Phosphate Max. 0.5 ppm
Aluminum Max. 0.2 ppm
Arsenic and Antimony Max. 0.005 ppm
Boron Max. 0.02 ppm
Calcium Max. 0.3 ppm
Chromium Max. 0.2 ppm
Copper Max. 0.1 ppm
Iron Max. 0.2 ppm
Lead Max. 0.3 ppm
Magnesium Max. 0.3 ppm
Manganese Max. 0.2 ppm
Nickel Max. 0.1 ppm
Potassium Max. 0.3 ppm
Sodium Max. 0.3 ppm
Tin Max. 0.2 ppm
Titanium Max. 0.3 ppm
Zinc Max. 0.2 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty