Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 25%, PharmaPUR®, GMP

Supplier: Puritan Products, Inc.
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

767615GMP 767655GMP 767605BGNGMP 767665GMP
76267-368EA 15103.08 USD InStock
76267-368 10061-062 10061-060 10061-058
Sodium hydroxide 25%, PharmaPUR®, GMP
Sodium hydroxide
Made from USP purified water and sodium hydroxide, NF pellets which also meet EP and JP specifications.

  • Custom concentration and packaging capabilities
  • GMP manufactured product
  • Full traceability and regulatory support

Used for a wide range pf applications include facility and equipment sanitization, pH adjusment of process streams, chromatography column cleaning, sanitization and storage.

The final solution is then filtered using a 0.2 μm filter.

Certifications: The solution meets all requirements for residual solvents USP <467>.


Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Appearance To Pass Test
Carbonate Max. 0.6%
Assay (w/w) 24.50 - 25.50%
Identification: Sodium Passes Test
Insoluble Substances and Organic Matter To Pass Test
Chloride Max. 30 ppm
Heavy Metals Max. 1 ppm
Iron Max. 1 ppm
Endotoxin (mL) 2.0 EU/mL maximum
Made from USP Purified Water and Sodium Hydroxide, NF pellets which also meet EP and JP specifications.
The final solution is then filtered using a 0.2µm filter. The solution meets all requirements for residual solvents USP <467>

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty