Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Pentyloxybenzaldehyde

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 4-Amyloxybenzaldehyde4-n-Amyloxybenzaldehyde4-(Pentyloxy)benzaldehyde4-n-Pentyloxybenzaldehyde

A1440-100GM
75871-734EA 227 USD InStock
75871-734
4-Pentyloxybenzaldehyde
4-Pentyloxybenzaldehyde


Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014135
CAS Number: 5736-91-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty