Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-6-ethoxybenzothiazole

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 6-Ethoxybenzothiazol-2-ylamine

A1442-25GM A1442-100GM
75871-736EA 282 USD InStock
75871-736 75871-738
2-Amino-6-ethoxybenzothiazole
2-Amino-6-ethoxybenzothiazole


Formula: C₉H₁₀N₂OS
Storage Temperature: Ambient
MDL Number: MFCD00005788
CAS Number: 94-45-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty