Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-5-chloro-2-methoxybenzoic acid

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 4-Amino-5-chloro-o-anisic acid

A1478-100GM A1478-25GM
75871-768EA 393 USD InStock
75871-768 75871-770
4-Amino-5-chloro-2-methoxybenzoic acid
4-Amino-5-chloro-2-methoxybenzoic acid


Formula: C₈H₈ClNO₃
MW: 201.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038794
CAS Number: 7206-70-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty