Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Pentylphenol

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 4-Amylphenol

A1582-25ML A1582-100ML
75871-804EA 147 USD InStock
75871-804 75871-778
4-Pentylphenol
4-Pentylphenol


Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00020211
CAS Number: 14938-35-3
UN: 3145
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty