Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alprazolam (CIV) USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 8-Chloro-1-methyl-6-phenyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepine

AL400-1GM AL400-50MG AL400-5GM AL400-1KGBL AL400-100MG AL400-100GMBL AL400-25MG AL400-25GMBL
76014-204EA 1846 USD InStock
76014-204 76014-214 76014-206 76014-200 76014-210 76014-202 76014-212 76014-208
Alprazolam (CIV) USP
Alprazolam
Alprazolam (CIV), USP is a short acting anxiolytic from the benzodiazepine class of drugs that are psychoactive. Alprazolam is a DEA CONTROLLED SUBSTANCE - SCHEDULE IV and does require DEA registration. All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Formula: C₁₇H₁₃ClN₄
MW: 308.76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078881
CAS Number: 28981-97-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty