Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(4-chlorobutyl)ether

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 4-Chlorobutyl Ether4,4'-Dichlorodibutyl Ether4,4'-Dichlorobutyl Ether

B1496-100ML B1496-25ML
75871-996EA 273 USD InStock
75871-996 75871-998
Bis(4-chlorobutyl)ether
Bis(4-chlorobutyl)ether


Formula: C₈H₁₆Cl₂O
MW: 199.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00019006
CAS Number: 6334-96-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty