Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2'-Bipyrimidyl, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 2,2'-Bipyrimidine2,2'-Dipyrimidyl

B1936-250MG B1936-1GM
75872-018EA 142 USD InStock
75872-018 75872-016
2,2'-Bipyrimidyl, Reagent Grade
2,2'-Bipyrimidyl


Formula: C₈H₆N₄
MW: 158.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014600
CAS Number: 34671-83-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty