Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Capsaicin USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 8-Methyl-N-vanillyl-6-none-amid

C1869-25GM C1869-1KGBL C1869-1GM C1869-100GMBL C1869-5GM
76014-302EA 354 USD InStock
76014-302 76014-304 76014-300 76014-306 76014-308
Capsaicin USP
Capsaicin
Capsaicin, USP is used as an analgesic in nasal spays, topical ointments and in dermal patches to relieve pain. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


Formula: C₁₈H₂₇NO₃
MW: 305.42 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00017259
CAS Number: 404-86-4
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty