Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopropyl methyl ketone (DEA List I Chemical)

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 1-Cyclopropylethanone

C2710-100ML C2710-25ML
75872-158EA 242 USD InStock
75872-158 75872-160
Cyclopropyl methyl ketone (DEA List I Chemical)
Cyclopropyl methyl ketone


Formula: C₅H₈O
MW: 84.12 g/mol
Density: 0.849
Storage Temperature: Ambient
MDL Number: MFCD00001297
CAS Number: 765-43-5
UN: 1224
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty