Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,1-Dimethylurea

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: N,N-Dimethylurea

D1428-25GM D1428-100GM
75872-264EA 129 USD InStock
75872-264 75872-262
1,1-Dimethylurea
1,1-Dimethylurea


Formula: C₃H₈N₂O
MW: 88.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007959
CAS Number: 598-94-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty