Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium-4,4'-bis(2-sulfonatostyryl)biphenyl

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Sodium 2,2'-([1,1'-biphenyl]-4,4'-diylbis(ethene-2,1-diyl))dibenzenesulphonate4,4'-Bis(2-sulfonatostyryl)biphenyl Disodium Salt4,4'-Bis(2-sulphonatostyryl)biphenyl Disodium Salt

D1608-100GM D1608-25GM
75872-320EA 93 USD InStock
75872-320 75872-318
Disodium-4,4'-bis(2-sulfonatostyryl)biphenyl
Disodium-4,4'-bis(2-sulfonatostyryl)biphenyl


Formula: C₂₈H₂₂O₆S₂
MW: 562.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00318874
CAS Number: 27344-41-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty