Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N'-Diphenyl-p-phenylenediamine

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: N,N'-Diphenyl-1,4-phenylenediamine

D1634-100GM D1634-25GM
75872-326EA 269 USD InStock
75872-326 75872-328
N,N'-Diphenyl-p-phenylenediamine
N,N'-Diphenyl-p-phenylenediamine


Formula: C₁₈H₁₆N₂
MW: 260.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003015
CAS Number: 74-31-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty