Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methyl-2-pentanone oxime

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Isobutyl Methyl KetoximeIsopropylacetone Oxime

M1760-100ML M1760-25ML
75872-984EA 131 USD InStock
75872-984 75872-982
4-Methyl-2-pentanone oxime
4-Methyl-2-pentanone oxime


Formula: C₆H₁₃NO
MW: 115.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013933
CAS Number: 105-44-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty