Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pregnenolone

Supplier: SPECTRUM CHEMICAL MFG CORP

P1422-100GM P1422-5GM P1422-1GM P1422-1KGBL P1422-25GM P1422-5KGBL P1422-25KGBL
75873-158EA 13413 USD InStock
75873-158 75873-148 75873-154 75873-156 76014-762 75873-150 75873-152
Pregnenolone
Pregnenolone
Pregnenolone | Non-graded Pregnenolone supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₂₁H₃₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00003628
CAS Number: 145-13-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty