Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pentadecanoic acid

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Pentadecylic acidn-Pentadecanoic acid

P2355-25GM P2355-5GM
75873-246EA 79 USD InStock
75873-246 75873-244
Pentadecanoic acid
Pentadecanoic acid
n-Pentadecanoic Acid, is a saturated fatty acid and is rare in nature but found in cows milk fat as well as in mutton fat that has been hydrogenated.


Formula: C₁₅H₃₀O₂
MW: 242.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002745
CAS Number: 1002-84-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty