Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Pyridylmethanol

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 3-PyridylmethanolPyridine-3-methanol3-Pyridinemethanol3-Pyridylcarbinol3-(Hydroxymethyl)pyridine

P2753-25ML P2753-100ML
75873-258EA 221 USD InStock
75873-258 75873-260
3-Pyridylmethanol
3-Pyridylmethanol
3-Pyridinemethanol is being investigated for its antineoplastic and vasodilator properties.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₆H₇NO
MW: 109.13 g/mol
Density: 1.124
Storage Temperature: Ambient
MDL Number: MFCD00006407
CAS Number: 100-55-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty