Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zirconium(IV) acetylacetonate

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Zirconium(IV) 2,4-pentanedionateTetrakis(2,4-pentanedionato)zirconium(IV)

T1578-25GM T1578-100GM
75873-420EA 95 USD InStock
75873-420 75873-418
Zirconium(IV) acetylacetonate
Zirconium (IV) acetylacetonate


Formula: C₂₀H₂₈O₈Zr
MW: 487.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000036
CAS Number: 17501-44-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty