Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2,2-Tribromoethanol

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Tribromoethyl alcohol

T1686-100GM T1686-25GM
75873-454EA 141.78 USD InStock
75873-454 75873-452
2,2,2-Tribromoethanol
2,2,2-Tribromoethanol


Formula: Br₃CCH₂OH
MW: 282.76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004671
CAS Number: 75-80-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty