Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ursodeoxycholic acid USP

Supplier: SPECTRUM CHEMICAL MFG CORP

U1304-25GM U1304-100GM U1304-5GM U1304-1KGBL U1304-5KGBL
76014-996EA 250 USD InStock
76014-996 76014-998 76014-992 76014-994 76014-990
Ursodeoxycholic acid USP
Ursodeoxycholic acid
Ursodiol, USP is a bile acid used to prevent and dissolve certain types of gallstones. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


Formula: C₂₄H₄₀O₄
MW: 392.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003680
CAS Number: 128-13-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty