Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rubber Bands, Anti-Static, Malaster Company

Supplier: Malaster Company
MCASRB10
16272-544BG 101.78 USD InStock
16272-544
Rubber Bands, Anti-Static, Malaster Company
Ties
These pink ESD rubber bands are made of durable vulcanized natural rubber. They are used to bundle and protect ESD sensitive IC tubes, trays, boxes and carriers quickly and safely.

  • Surface resistivity: 10e7 to 10e9 ohms/sq
  • Static decay rate: Less than two seconds
  • ESD shelf life: Approximately one year
  • Stretch up to 550% elongation
  • Tensile strength is minimum 12Mp
  • Heavy metal free

The rubber bands are recyclable and are packed approximately 175 per bag (1 kg).

Certifications: RoHS Compliant.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty