Would you like to log in again?
ChiraDex HPLC Columns, Agilent Technologies
Supplier: Agilent Technologies
79925CB-584 79925CB-584
AG79925CB-584EA 2194 USD InStock
AG79925CB-584 CA79925CB-584
ChiraDex HPLC Columns, Agilent Technologies
Chromatography Columns
ChiraDex columns allow for direct racemic separations without derivatization.
  • Cartridge columns
  • Novel manufacturing process bonds ß-cyclodextrin to spherical 5 µm silica gel by means of a chemical spacer
Applications include barbiturates, benzothiadiazines, calcium antagonists, and PTH-amino acids.
Note: Cartridge columns require reusable cartridge hardware (101481-418).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty