Would you like to log in again?
Versaflo™ Healthcare Powered Air Purifying Respirator (PAPR) Kit TR-300N+ HKS, 3M™
Supplier: 3M
Versaflo™
7100153808
76220-738CS 2002.03 USD InStock
76220-738
Versaflo™ Healthcare Powered Air Purifying Respirator (PAPR) Kit TR-300N+ HKS, 3M™
Respirators Respiratory Systems
3M™ Versaflo™ TR-300N+ HKS is a complete, ready-to-use HE PAPR system with a small-to-medium headcover. Suggested industries include, pharmaceutical and healthcare. Provides protection against particulates.
  • Easy access to ears
  • Wider field of vision with enhanced visor design
  • Conveniently disposable with maintenance-free disposable hood
  • Two selectable flow rate options - 6.5 CFM and 7.2 CFM
  • Easy-to-use and versatile PAPR system for particulate environments
  • High efficiency particulate filter
  • Rechargeable high capacity lithium ion battery
Stethoscope use is easier with the unique shape of our S-Series headcovers. The innovative Quick Release Swivel (QRS) connects quickly and helps avoid kinks in the breathing tube. Two types of alarms one audible and one visual, alert user to either a low batter or low flow, such as when a particle filter is fully loaded. LED lights on the battery indicate its charge status on and off the charger.
Certifications: NIOSH approved.
Delivery information: The kit includes, headcover, length adjusting breathing tube with breathing tube cover, carrying bag, PAPR unit, easy clean belt, high capacity battery, battery charger kit, HE filters, prefilter, airflow indicator.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty