Would you like to log in again?
Hydrion® Brilliant Dip-Stik Plastic Strip, Micro Essential Laboratory®
Supplier: Micro Essential Laboratory
7000
89370-902CS 179.11 USD InStock
89370-902
Hydrion® Brilliant Dip-Stik Plastic Strip, Micro Essential Laboratory®
Test Paper pH Indicator Papers and Test Strips
This Insta-Chek pH test dip-stik provides excellent color separation. These wide-range dip-stiks offer clear bright single color matches at every 1.0 interval from pH 0.0 - 13.0.
The color chart has pH matches at [0.0][1.0][2.0][3.0][4.0][5.0][6.0][7.0][8.0][9.0][10.0][11.0][12.0][13.0].
Delivery information: Each carton contains 10 cases with 100 strips per case. Provides for approximately 1000 tests.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty