Would you like to log in again?
Bovine Ribonuclease A (from Pancreas)
Supplier: EMD Millipore
Novagen®
70856-3
EM70856-3EA 125.31 USD InStock
EM70856-3
Bovine Ribonuclease A (from Pancreas)
Enzymes
Convenient solution for selective degradation of RNA.
This is a convenient alternative to powdered RNase A. It is a highly purified preparation of bovine pancreatic ribonuclease A suitable for use in selective removal of RNA. It has been pretreated to remove DNase I and is suitable for RNA digestion in plasmid purification procedures.
Delivery information: Supplied at a concentration of 10 mg/ml in 10 mM Tris-HCl, 1 mM EDTA, 50% glycerol, pH 7.5.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty