Would you like to log in again?
SterEM™ Sabdex Agar with Lecithin and Tween® 80, USP, Irradiated, Hardy Diagnostics
Supplier: Hardy Diagnostics
W595
76200-290PK 82.36 USD InStock
76200-290
SterEM™ Sabdex Agar with Lecithin and Tween® 80, USP, Irradiated, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Sterile plates for environmental monitoring.
  • Room temperature storage
  • Improved moisture control packaging format
  • Optically clear polystyrene - For distortion free viewing
  • Heavy-Duty - Reduces breakage during shipping
  • Stacking rings - Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
  • Sterile by Irradiation - Validated for Sterility Assurance Level (SAL) of 10-5
  • Triple Bagged - Two nylon inner bags. One outer Mylar(R) bag that is heat sealed
  • Outer bag is VHP impermeable
  • Certificate of Analysis - Available at the Hardy Diagnostics website 24/7
  • Manufactured in a licensed cGMP facility with a quality management system certified to ISO 13485
Irradiated Sabouraud Dextrose (SabDex) agar with lecithin and Tween® 80 is recommended for the detection and enumeration of yeasts and fungi from environmental sources in a clean room setting. The plates are triple bagged and sterilized by irradiation to promote a higher sterility assurance level.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty