Would you like to log in again?
J&W CP-Wax 58 FFAP CB GC Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
CP7717I5 CP7665 CP7614 CP7727 CP7767 CP7717 CP7778 CP7727 CP7747 CP7797 CP7624 CP7654 CP7787 CP7757 CP7777 CP7664
AGCP7727EA 1098 USD InStock
CAAGCP7727 AGCP7727 101475-182 101406-534 101406-632 103578-220 101475-118 101475-228 101475-204 101475-220 101406-532 101406-630 101406-520 101475-096 101475-222 101475-156
J&W CP-Wax 58 FFAP CB GC Columns, Agilent Technologies
Chromatography Columns
Agilent JW CP-Wax 58 FFAP CB column is a nitroterephthalic acid-modified, chemically bonded polyethylene glycol phase.
  • Highest polarity bonded wax column for analyzing polar compounds
CP-Wax 58 FFAP is designed for acidic compounds such as phenols and free fatty acids, both derivatized and underivatized.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty