Would you like to log in again?
Premium 3-Layer Vinyl Worksurface Mats, Desco
Supplier: Desco Industries
TM24600L3BL TM36600L3BL TM30600L3BL
89024-576EA 648.9 USD InStock
89024-576 89024-580 89024-584
Premium 3-Layer Vinyl Worksurface Mats, Desco
Surface Protectors Bench Protectors
This three-layer vinyl material provides excellent electrical properties.
  • Made from a tough, easy-to-clean dissipative vinyl
  • Available in rolls, mats, and mats with a ground cord
The center layer, a highly conductive metal scrim, moves the charge from the surface to ground quickly. The bottom layer is a durable, non-skid, cushion of dissipative foam. Compatible with constant monitors.
Certifications: Meets ANSI/ESD S20.20 worksurface required limits and recommendation of ANSI/ESD S4.1.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty