Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Type 2 HEPA Filter for Toner and Dust, SCS, Desco Industries

Supplier: Desco Industries

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

SV-MPF2
89204-681EA 38.55 USD InStock
89204-681
Type 2 HEPA Filter for Toner and Dust, SCS, Desco Industries
Filters Gas Filters
Specially designed for trapping unwanted toner from copiers and laser printers. This filter is also ideal for cleaning keyboards, fans, and other household dust collections.

  • Used for dust or dry pick-up
  • Efficiency: 99%
  • Size range: 3.0 to 5.0 µm
  • CFM: 40 cubic feet per minute
  • Compatible with 497ABG, 497AJN, 497AJX

This replaceable pleated filter is encased in plastic, making it durable and easy to dispose.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty