Would you like to log in again?
Cow Longitudinal Section of Bone
Supplier: Ward's Science
140.1776.1
470220-630EA 29.45 USD InStock
470220-630
Cow Longitudinal Section of Bone
Models Zoology Models
Cow longitudinal secton of bone.
Front leg bone to view internal structure, periosteum, solid bone layer, spongy bone, medullary canal, yellow bone marrow, red bone marrow, epiphysial line, head of the long bone with articulating surface of the hyaline cartilage.
Preserved cow material is a byproduct of the food industry. Cows are not raised specifically for dissection. The animal material is “recycled” for educational use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty