Would you like to log in again?
MicroMax® NS Disposable Boot Covers, Lakeland Industries
Supplier: Lakeland Industries
CTL903P-XL CTL903P-2X
76332-678CS 487.47 USD InStock
76332-678 76332-676
MicroMax® NS Disposable Boot Covers, Lakeland Industries
Disposable Garments
Disposable protective boot covers are made from lightweight microporous fabric for protection from bloodborne pathogens.
  • Microporous film laminated to polypropylene substrate
  • Serged seams
  • Blood-borne pathogen/viral barrier tested
  • Elastic closure at the top
These boot covers provide the protection needed from dirt, dust, grease, grime, and light chemical splash, as well as blood/viral protection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty