Would you like to log in again?
MicroMax® NS Disposable Protective Sleeves, Lakeland Industries
Supplier: Lakeland Industries
CTL850P-18
76332-594CS 308.98 USD InStock
76332-594
MicroMax® NS Disposable Protective Sleeves, Lakeland Industries
Disposable Garments
Disposable protective sleeves are made from lightweight microporous fabric for protection from bloodborne pathogens.
  • Microporous film laminated to polypropylene substrate
  • Serged seams
  • Blood-borne pathogen/viral barrier tested
  • Elastic closure at each end to provide protection from the wrist to above the elbow
These sleves provide the protection needed from dirt, dust, grease, grime, and light chemical splash, as well as blood/viral protection.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty